Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Propylbiphenyl-4-carboxylic acid, 98%

$48.34 - $160.08

Chemical Identifiers

Chemical Name or Material 4'-n-Propylbiphenyl-4-carboxylic acid
CAS 88038-94-2
Molecular Formula C16H16O2
Formula Weight 240.3
MDL Number MFCD00017335
Beilstein 6314193
Synonym 4-(4-n-Propylphenyl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0611003

 
alfa aesar™
L0611003
1g Each for $48.34

AAL0611006

 
alfa aesar™
L0611006
5g Each for $160.08
Description & Specifications

Specifications

Formula Weight 240.3
Chemical Name or Material 4'-n-Propylbiphenyl-4-carboxylic acid
Assay 98%
Molecular Formula C16H16O2
MDL Number MFCD00017335
Synonym 4-(4-n-Propylphenyl)benzoic acid
Quantity 1g
CAS 88038-94-2
Beilstein 6314193
Melting Point 224° to 228°C