Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propylbenzoic acid, 98+%

$47.16 - $2,463.55

Chemical Identifiers

Chemical Name or Material 4-n-Propylbenzoic acid
CAS 2438-05-3
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00013996
Beilstein 1934550
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1453809

  SDS
alfa aesar™
A1453809
10g Each for $47.16

AAA1453830

  SDS
alfa aesar™
A1453830
250g Each for $2,463.55
Description & Specifications

Specifications

Formula Weight 164.2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 1934550
Assay ≥98%
Chemical Name or Material 4-n-Propylbenzoic acid
Melting Point 139° to 144°C
Molecular Formula C10H12O2
CAS 2438-05-3
Quantity 10g
Health Hazard 1 Warning
MDL Number MFCD00013996
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.