missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propylbenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1453818

 View more versions of this product

Catalog No. AAA1453818



Specifications

Specifications

4-n-Propylbenzoic acid
50g
C10H11O2
1934550
ATZHGRNFEFVDDJ-UHFFFAOYSA-M
4-propylbenzoic acid
137601
≥98%
139°C to 144°C
2438-05-3
MFCD00013996
4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348
CCCC1=CC=C(C=C1)C(=O)O
163.20
164.2

Chemical Identifiers

2438-05-3
163.20
ATZHGRNFEFVDDJ-UHFFFAOYSA-M
137601
CCCC1=CC=C(C=C1)C(=O)O
C10H11O2
MFCD00013996
4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348
4-propylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents