Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propylbenzenesulfonyl chloride, 95%

Chemical Identifiers

Chemical Name or Material 4-n-Propylbenzenesulfonyl chloride
CAS 146949-07-7
Molecular Formula C9H11ClO2S
Formula Weight 218.7
MDL Number MFCD00041508
UN Number UN3265
Beilstein 5511216
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1835406

  SDS
alfa aesar™
A1835406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 218.7
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 5511216
Chemical Name or Material 4-n-Propylbenzenesulfonyl chloride
CAS 146949-07-7
Melting Point 20° to 22°C
Boiling Point 275° to 276°C
Molecular Formula C9H11ClO2S
UN Number UN3265
Assay 95%
Quantity 5g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
MDL Number MFCD00041508
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.