Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propylaniline, 98%

Chemical Identifiers

Chemical Name or Material 4-n-Propylaniline
CAS 2696-84-6
Molecular Formula C9H13N
Formula Weight 135.21
MDL Number MFCD00007924
UN Number UN2810
Beilstein 2205524
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0388003

 
alfa aesar™
L0388003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0388006

 
alfa aesar™
L0388006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0388014

 
alfa aesar™
L0388014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.21
TSCA TSCA
Assay 98%
Molecular Formula C9H13N
MDL Number MFCD00007924
CAS 2696-84-6
UN Number UN2810
Beilstein 2205524
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECS Number BY8320000
Quantity 1g
Chemical Name or Material 4-n-Propylaniline
Flash Point 103°C (217°F)
Boiling Point 224° to 226°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.