missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Propylacetophenone, 97%

$98.84 - $313.05

Chemical Identifiers

CAS 2932-65-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00041359
InChI Key ZNBVIYMIVFKTIW-UHFFFAOYSA-N
Synonym p-propylacetophenone, 1-4-propylphenyl ethan-1-one, 1-4-propylphenyl ethanone, 4'-propylacetophenone, ethanone, 1-4-propylphenyl, 4-n-propylacetophenone, 4'-n-propylacetophenone, 1-acetyl-4-propylbenzene, p-propylacetophenon, p-n-propylacetophenone
PubChem CID 76236
IUPAC Name 1-(4-propylphenyl)ethanone
SMILES CCCC1=CC=C(C=C1)C(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0114414 View Documents Alfa Aesar™
L0114414
25g Each for $98.84
 
AAL0114422 View Documents Alfa Aesar™
L0114422
100g Each for $313.05
 
Specifications

Chemical Identifiers

2932-65-2
162.232
ZNBVIYMIVFKTIW-UHFFFAOYSA-N
76236
CCCC1=CC=C(C=C1)C(=O)C
C11H14O
MFCD00041359
p-propylacetophenone, 1-4-propylphenyl ethan-1-one, 1-4-propylphenyl ethanone, 4'-propylacetophenone, ethanone, 1-4-propylphenyl, 4-n-propylacetophenone, 4'-n-propylacetophenone, 1-acetyl-4-propylbenzene, p-propylacetophenon, p-n-propylacetophenone
1-(4-propylphenyl)ethanone

Specifications

251°C to 252°C
2932-65-2
MFCD00041359
p-propylacetophenone, 1-4-propylphenyl ethan-1-one, 1-4-propylphenyl ethanone, 4'-propylacetophenone, ethanone, 1-4-propylphenyl, 4-n-propylacetophenone, 4'-n-propylacetophenone, 1-acetyl-4-propylbenzene, p-propylacetophenon, p-n-propylacetophenone
CCCC1=CC=C(C=C1)C(=O)C
162.232
162.23
4'-n-Propylacetophenone
25g
C11H14O
1858604
ZNBVIYMIVFKTIW-UHFFFAOYSA-N
1-(4-propylphenyl)ethanone
76236
97%