Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propyl-4-heptanol, 99%

$116.68 - $116.68

Chemical Identifiers

Chemical Name or Material 4-n-Propyl-4-heptanol
CAS 2198-72-3
Molecular Formula C4H22O
Linear Formula (CH3CH2CH2)3COH
Formula Weight 158.28
MDL Number MFCD00046604
Synonym Tri-n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1994406

 
alfa aesar™
A1994406
5g Each for $116.68
Description & Specifications

Specifications

Formula Weight 158.28
Chemical Name or Material 4-n-Propyl-4-heptanol
Assay 99%
Molecular Formula C4H22O
Linear Formula (CH3CH2CH2)3COH
MDL Number MFCD00046604
CAS 2198-72-3
Synonym Tri-n-propyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.