Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propyl-1-hepten-4-ol, 98%

$84.04 - $1,077.50

Chemical Identifiers

Chemical Name or Material 4-n-Propyl-1-hepten-4-ol
CAS 62108-07-0
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00049206
Synonym Allyl di-n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2042906

 
alfa aesar™
B2042906
5g Each for $84.04

AAB2042914

 
alfa aesar™
B2042914
25g Each for $330.95

AAB2042922

 
alfa aesar™
B2042922
100g Each for $1,077.50
Description & Specifications

Specifications

Formula Weight 156.27
Chemical Name or Material 4-n-Propyl-1-hepten-4-ol
Assay 98%
Molecular Formula C10H20O
MDL Number MFCD00049206
CAS 62108-07-0
Synonym Allyl di-n-propyl carbinol
Quantity 5g