Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Propoxycarbonyl)benzeneboronic acid, 97%

$59.38 - $176.30

Chemical Identifiers

Chemical Name or Material 4-(n-Propoxycarbonyl)benzeneboronic acid
CAS 91062-38-3
Molecular Formula C10H13BO4
Formula Weight 208.02
MDL Number MFCD06659852
Synonym 4-(n-Propoxycarbonyl)phenylboronic acid; n-Propyl 4-boronobenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2793803

  SDS
alfa aesar™
H2793803
1g Each for $59.38

AAH2793809

  SDS
alfa aesar™
H2793809
10g Each for $176.30
Description & Specifications

Specifications

Formula Weight 208.02
Quantity 1g
Molecular Formula C10H13BO4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 121° to 123°C
Chemical Name or Material 4-(n-Propoxycarbonyl)benzeneboronic acid
Synonym 4-(n-Propoxycarbonyl)phenylboronic acid; n-Propyl 4-boronobenzoate
MDL Number MFCD06659852
CAS 91062-38-3
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.