Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Propoxybiphenyl-4-boronic acid, 97%

Chemical Identifiers

Chemical Name or Material 4'-n-Propoxybiphenyl-4-boronic acid
CAS 849062-20-0
Formula Weight 256.11
MDL Number MFCD06798061
Synonym 4-(4'-Propoxyphenyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5599903

 
alfa aesar™
H5599903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55999MD

 
alfa aesar™
H55999MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.11
CAS 849062-20-0
Synonym 4-(4'-Propoxyphenyl)phenylboronic acid
Assay 97%
MDL Number MFCD06798061
Quantity 1g
Physical Form Solid
Chemical Name or Material 4'-n-Propoxybiphenyl-4-boronic acid