missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propoxybenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5312906

 View more versions of this product

Catalog No. AAH5312906



Specifications

Specifications

4-n-Propoxybenzeneboronic acid
5g
C9H13BO3
4-propoxybenzeneboronic acid, 4-n-propoxyphenylboronic acid, 4-propoxyphenyl boronic acid, 4-propyloxyphenyl boronic acid, 4-n-propoxybenzeneboronic acid, p-propoxyphenylboronic acid, 4-propoxyphenylboronicacid, boronic acid, 4-propoxyphenyl, b-4-propoxyphenyl boronic acid, pubchem9546
B(C1=CC=C(C=C1)OCCC)(O)O
180.01
180.01
104°C to 110°C
186497-67-6
MFCD03427189
SOKFEJKZYHYKSY-UHFFFAOYSA-N
(4-propoxyphenyl)boronic acid
4456490
97%

Chemical Identifiers

186497-67-6
180.01
SOKFEJKZYHYKSY-UHFFFAOYSA-N
4456490
B(C1=CC=C(C=C1)OCCC)(O)O
C9H13BO3
MFCD03427189
4-propoxybenzeneboronic acid, 4-n-propoxyphenylboronic acid, 4-propoxyphenyl boronic acid, 4-propyloxyphenyl boronic acid, 4-n-propoxybenzeneboronic acid, p-propoxyphenylboronic acid, 4-propoxyphenylboronicacid, boronic acid, 4-propoxyphenyl, b-4-propoxyphenyl boronic acid, pubchem9546
(4-propoxyphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents