missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Propoxyaniline, 97%

Manufacturer:  Alfa Aesar™ H5509606

 View more versions of this product

Catalog No. AAH5509606



Description

Description

Applications
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.

Solubility
Slightly soluble (1.3 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-n-Propoxyaniline
5g
C9H13NO
4-n-propoxyaniline
CCCOC1=CC=C(C=C1)N
151.209
151.21
97%
Brown
4469-80-1
MFCD00043621
DWOIGSLSPPLRKO-UHFFFAOYSA-N
4-propoxyaniline
78221
Liquid

Chemical Identifiers

4469-80-1
151.209
DWOIGSLSPPLRKO-UHFFFAOYSA-N
78221
CCCOC1=CC=C(C=C1)N
C9H13NO
MFCD00043621
4-n-propoxyaniline
4-propoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents