missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentylphenyl 4-n-propylbenzoate, 97%

Manufacturer:  Alfa Aesar™ H5530606

 View more versions of this product

Catalog No. AAH5530606



Specifications

Specifications

4-n-Pentylphenyl 4-n-propylbenzoate
1.543
5g
C21H26O2
4-n-pentylphenyl 4-n-propylbenzoate
CCCCCC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)CCC
310.437
310.44
0.989
15°C to 18°C
50649-60-0
MFCD00041941
WNBFPAKRCJNBBS-UHFFFAOYSA-N
(4-pentylphenyl) 4-propylbenzoate
170879
97%

Chemical Identifiers

50649-60-0
310.437
WNBFPAKRCJNBBS-UHFFFAOYSA-N
170879
CCCCCC1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)CCC
C21H26O2
MFCD00041941
4-n-pentylphenyl 4-n-propylbenzoate
(4-pentylphenyl) 4-propylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

EINECSNumber : 256-684-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents