Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentyloxybenzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-n-Pentyloxybenzoyl chloride
CAS 36823-84-4
Molecular Formula C12H15ClO2
Formula Weight 226.70g/mol
MDL Number MFCD00041723
UN Number UN3265
Beilstein 2937454
Synonym 4-n-Amyloxybenzoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1501903

 
alfa aesar™
A15019-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1501906

 
alfa aesar™
A15019-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1501914

 
alfa aesar™
A15019-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 198° to 200°C (30mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Chemical Name or Material 4-n-Pentyloxybenzoyl chloride
CAS 36823-84-4
Assay 97%
Molecular Formula C12H15ClO2
Formula Weight 226.70g/mol
MDL Number MFCD00041723
UN Number UN3265
Beilstein 2937454
Synonym 4-n-Amyloxybenzoyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.