Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentyloxybenzoic acid, 98%

$53.73 - $184.72

Chemical Identifiers

Chemical Name or Material 4-n-Pentyloxybenzoic acid
CAS 15872-41-0
Molecular Formula C12H16O3
Formula Weight 208.26
MDL Number MFCD00013990
Beilstein 1951836
Synonym 4-n-Amyloxybenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1502406

 
alfa aesar™
A1502406
5g Each for $53.73

AAA1502414

 
alfa aesar™
A1502414
25g Each for $184.72
Description & Specifications

Specifications

Formula Weight 208.26
Melting Point 123°C (N)
Chemical Name or Material 4-n-Pentyloxybenzoic acid
TSCA TSCA
Assay 98%
Molecular Formula C12H16O3
MDL Number MFCD00013990
CAS 15872-41-0
Beilstein 1951836
Synonym 4-n-Amyloxybenzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.