Learn More
Alfa Aesar™ 4-n-Pentyloxybenzoic acid, 98%
Manufacturer: Alfa Aesar™ A1502414
Specifications
4-n-Pentyloxybenzoic acid | |
25g | |
C12H16O3 | |
1951836 | |
OZPPUPJQRJYTNY-UHFFFAOYSA-N | |
4-pentoxybenzoic acid | |
85153 | |
98% |
123°C (N) | |
15872-41-0 | |
MFCD00013990 | |
4-pentyloxybenzoic acid, 4-pentyloxy benzoic acid, 4-n-pentyloxybenzoic acid, benzoic acid, 4-pentyloxy, p-pentyloxybenzoic acid, 4-n-amyloxybenzoic acid, p-pentoxybenzoic acid, p-n-pentyloxybenzoic acid, 4-amyloxybenzoic acid, p-pentyloxy benzoic acid | |
CCCCCOC1=CC=C(C=C1)C(=O)O | |
208.257 | |
208.26 |
Chemical Identifiers
15872-41-0 | |
208.257 | |
OZPPUPJQRJYTNY-UHFFFAOYSA-N | |
85153 | |
CCCCCOC1=CC=C(C=C1)C(=O)O |
C12H16O3 | |
MFCD00013990 | |
4-pentyloxybenzoic acid, 4-pentyloxy benzoic acid, 4-n-pentyloxybenzoic acid, benzoic acid, 4-pentyloxy, p-pentyloxybenzoic acid, 4-n-amyloxybenzoic acid, p-pentoxybenzoic acid, p-n-pentyloxybenzoic acid, 4-amyloxybenzoic acid, p-pentyloxy benzoic acid | |
4-pentoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA