missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentyloxybenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1502406

 View more versions of this product

Catalog No. AAA1502406



Specifications

Specifications

4-n-Pentyloxybenzoic acid
5g
C12H16O3
1951836
OZPPUPJQRJYTNY-UHFFFAOYSA-N
4-pentoxybenzoic acid
85153
98%
123°C (N)
15872-41-0
MFCD00013990
4-pentyloxybenzoic acid, 4-pentyloxy benzoic acid, 4-n-pentyloxybenzoic acid, benzoic acid, 4-pentyloxy, p-pentyloxybenzoic acid, 4-n-amyloxybenzoic acid, p-pentoxybenzoic acid, p-n-pentyloxybenzoic acid, 4-amyloxybenzoic acid, p-pentyloxy benzoic acid
CCCCCOC1=CC=C(C=C1)C(=O)O
208.257
208.26

Chemical Identifiers

15872-41-0
208.257
OZPPUPJQRJYTNY-UHFFFAOYSA-N
85153
CCCCCOC1=CC=C(C=C1)C(=O)O
C12H16O3
MFCD00013990
4-pentyloxybenzoic acid, 4-pentyloxy benzoic acid, 4-n-pentyloxybenzoic acid, benzoic acid, 4-pentyloxy, p-pentyloxybenzoic acid, 4-n-amyloxybenzoic acid, p-pentoxybenzoic acid, p-n-pentyloxybenzoic acid, 4-amyloxybenzoic acid, p-pentyloxy benzoic acid
4-pentoxybenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents