Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentyloxybenzaldehyde, 97%

$67.58 - $252.45

Chemical Identifiers

Chemical Name or Material 4-n-Pentyloxybenzaldehyde
CAS 5736-91-4
Molecular Formula C12H16O2
Formula Weight 192.26
MDL Number MFCD00014135
Beilstein 642040
Synonym 4-n-Amyloxybenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0908914

  SDS
alfa aesar™
L0908914
25g Each for $252.45

AAL0908906

  SDS
alfa aesar™
L0908906
5g Each for $67.58
Description & Specifications

Specifications

Formula Weight 192.26
Chemical Name or Material 4-n-Pentyloxybenzaldehyde
MDL Number MFCD00014135
CAS 5736-91-4
TSCA TSCA
Assay 97%
Molecular Formula C12H16O2
Beilstein 642040
Synonym 4-n-Amyloxybenzaldehyde
Boiling Point 144° to 147°C (1mmHg)
Quantity 25g