Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentylcyclohexanone, 98+%

Chemical Identifiers

Chemical Name or Material 4-n-Pentylcyclohexanone
CAS 61203-83-6
Molecular Formula C11H20O
Formula Weight 168.28
MDL Number MFCD00059581
UN Number UN3082
Synonym 4-n-Amylcyclohexanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2208818

  SDS
alfa aesar™
B2208818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.28
Chemical Name or Material 4-n-Pentylcyclohexanone
Assay ≥98%
Molecular Formula C11H20O
MDL Number MFCD00059581
CAS 61203-83-6
UN Number UN3082
Synonym 4-n-Amylcyclohexanone
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Quantity 50g