Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentylbiphenyl, 99%

$106.99 - $344.80

Chemical Identifiers

Chemical Name or Material 4-n-Pentylbiphenyl
CAS 7116-96-3
Molecular Formula C17H20
Formula Weight 224.35
MDL Number MFCD00009503
Beilstein 2082056
Synonym 4-n-Amylbiphenyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1259006

 
alfa aesar™
A1259006
5g Each for $106.99

AAA1259014

 
alfa aesar™
A1259014
25g Each for $344.80
Description & Specifications

Specifications

Formula Weight 224.35
Beilstein 2082056
Synonym 4-n-Amylbiphenyl
Chemical Name or Material 4-n-Pentylbiphenyl
TSCA TSCA
Assay 99%
Molecular Formula C17H20
MDL Number MFCD00009503
CAS 7116-96-3
Flash Point >110°C (230°F)
Boiling Point 166° to 167°C (6mmHg)
Melting Point 9° to 11°C
Quantity 5g