missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentylbenzoyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1024806

 View more versions of this product

Catalog No. AAA1024806



Description

Description

Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
Specifications

Specifications

4-n-Pentylbenzoyl chloride
144°C to 145°C (10mmHg)
49763-65-7
MFCD00000700
2207887
FBBRKYLXMNQFQU-UHFFFAOYSA-N
4-pentylbenzoyl chloride
170812
98%
>110°C (230°F)
5g
C12H15ClO
UN3265
benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride
CCCCCC1=CC=C(C=C1)C(=O)Cl
210.701
210.71

Chemical Identifiers

49763-65-7
210.701
FBBRKYLXMNQFQU-UHFFFAOYSA-N
170812
CCCCCC1=CC=C(C=C1)C(=O)Cl
C12H15ClO
MFCD00000700
benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride
4-pentylbenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents