Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentylbenzoic acid, 99%

$80.61 - $80.61

Chemical Identifiers

Chemical Name or Material 4-n-Pentylbenzoic acid
CAS 26311-45-5
Molecular Formula C12H16O2
Formula Weight 192.26
MDL Number MFCD00002572
Beilstein 2044798
Synonym 4-n-Amylbenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1208606

  SDS
alfa aesar™
A1208606
5g Each for $80.61

AAA1208614

  SDS
alfa aesar™
A1208614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-n-Pentylbenzoic acid
Quantity 5g
CAS 26311-45-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DH6273000
TSCA TSCA
Assay 99%
Molecular Formula C12H16O2
Formula Weight 192.26
MDL Number MFCD00002572
Beilstein 2044798
Synonym 4-n-Amylbenzoic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.