Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Pentylbenzeneboronic acid, 97%

$87.28 - $960.51

Chemical Identifiers

Chemical Name or Material 4-n-Pentylbenzeneboronic acid
CAS 121219-12-3
Molecular Formula C11H17BO2
Formula Weight 192.07
MDL Number MFCD00995151
Beilstein 4782754
Synonym 4-n-Amylbenzeneboronic acid; 4-n-Pentylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1801103

 
alfa aesar™
L1801103
1g Each for $252.45

AAL1801106

 
alfa aesar™
L1801106
5g Each for $960.51

AAL18011MD

 
alfa aesar™
L18011MD
250mg Each for $87.28
Description & Specifications

Specifications

Formula Weight 192.07
Chemical Name or Material 4-n-Pentylbenzeneboronic acid
CAS 121219-12-3
Assay 97%
Molecular Formula C11H17BO2
MDL Number MFCD00995151
Beilstein 4782754
Synonym 4-n-Amylbenzeneboronic acid; 4-n-Pentylphenylboronic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 82° to 84°C
Quantity 1g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.