Learn More
4-n-Pentylaniline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2656914
Description
4-Pentylaniline is used in the synthesis of 4-[(4?-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Pentylaniline is used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol
Solubility
Sparingly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
4-n-Pentylaniline | |
0.919 | |
>110°C (230°F) | |
1.53 | |
25 g | |
Sparingly soluble in water. | |
CCCCCC1=CC=C(C=C1)N | |
163.264 | |
163.26 |
33228-44-3 | |
130°C | |
C11H17N | |
MFCD00007926 | |
benzenamine, 4-pentyl, 4-amylaniline, p-pentylaniline, p-amylaniline, unii-8roj0g9hiu, p-n-pentylaniline, 4-n-pentylaniline, 4-pentylphenylamine, 8roj0g9hiu, p-n-amylaniline | |
DGFTWBUZRHAHTH-UHFFFAOYSA-N | |
4-pentylaniline | |
93162 | |
98% |
Chemical Identifiers
33228-44-3 | |
163.264 | |
DGFTWBUZRHAHTH-UHFFFAOYSA-N | |
93162 | |
CCCCCC1=CC=C(C=C1)N |
C11H17N | |
MFCD00007926 | |
benzenamine, 4-pentyl, 4-amylaniline, p-pentylaniline, p-amylaniline, unii-8roj0g9hiu, p-n-pentylaniline, 4-n-pentylaniline, 4-pentylphenylamine, 8roj0g9hiu, p-n-amylaniline | |
4-pentylaniline |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 251-409-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only