Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Pentylacetophenone, 98%

$201.65 - $201.65

Chemical Identifiers

Chemical Name or Material 4'-n-Pentylacetophenone
CAS 37593-02-5
Molecular Formula C13H18O
Formula Weight 190.29
MDL Number MFCD00043687
Synonym 4'-n-Amylacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1072506

 
alfa aesar™
A1072506
5g Each for $201.65
Description & Specifications

Specifications

Formula Weight 190.29
Chemical Name or Material 4'-n-Pentylacetophenone
TSCA TSCA
Assay 98%
Molecular Formula C13H18O
MDL Number MFCD00043687
CAS 37593-02-5
Synonym 4'-n-Amylacetophenone
Boiling Point 132° to 133°C (4mmHg)
Quantity 5g