Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Octyloxybenzoic acid, 98%

$62.23 - $62.23

Chemical Identifiers

Chemical Name or Material 4-n-Octyloxybenzoic acid
CAS 2493-84-7
Molecular Formula C15H22O3
Formula Weight 250.34
MDL Number MFCD00013993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2265909

  SDS
alfa aesar™
B2265909
10g Each for $62.23
Description & Specifications

Specifications

Formula Weight 250.34
TSCA TSCA
MDL Number MFCD00013993
Chemical Name or Material 4-n-Octyloxybenzoic acid
Molecular Formula C15H22O3
Quantity 10g
Assay 98%
CAS 2493-84-7
Melting Point 101° to 105°C