Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Octyloxyacetophenone

$207.80 - $920.49

Chemical Identifiers

Chemical Name or Material 4'-n-Octyloxyacetophenone
CAS 37062-63-8
Molecular Formula C16H24O2
Formula Weight 248.37
MDL Number MFCD00143208
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5068403

 
alfa aesar™
H5068403
1g Each for $207.80

AAH5068406

 
alfa aesar™
H5068406
5g Each for $920.49
Description & Specifications

Specifications

Formula Weight 248.37
Chemical Name or Material 4'-n-Octyloxyacetophenone
Quantity 1g
Molecular Formula C16H24O2
MDL Number MFCD00143208
CAS 37062-63-8