missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(n-Octyloxy)phenol, 98%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 352840010

 View more versions of this product

Catalog No. AC352840010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

222.33
100.0
98%
98%
3780-50-5
1g
97.5
4-(n-Octyloxy)phenol
C14H22O2

Chemical Identifiers

3780-50-5
100.0
C14H22O2
4-(n-Octyloxy)phenol, 98%
97.5
98%
222.33

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 223-243-6

TSCA : TSCA