missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Octylbiphenyl, 97%

$100.84 - $1,434.58

Chemical Identifiers

CAS 7116-97-4
Molecular Formula C20H26
Formula Weight 266.43
MDL Number MFCD00102113
Chemical Name or Material 4-n-Octylbiphenyl
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2501003 SDS Alfa Aesar™
B2501003
1g Each for $100.84
AAB2501006 SDS Alfa Aesar™
B2501006
5g Each for $353.04
AAB2501014 SDS Alfa Aesar™
B2501014
25g Each for $1,434.58
Specifications Description & Specifications

Chemical Identifiers

7116-97-4
266.43
4-n-Octylbiphenyl
C20H26
MFCD00102113

Specifications

7116-97-4
C20H26
MFCD00102113
97%
1g
266.43
4-n-Octylbiphenyl