missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-n-Octylacetophenone 97.0+%, TCI America™

Manufacturer:  TCI America O020725G

 View more versions of this product

Catalog No. O020725G



Specifications

Specifications

4′-n-Octylacetophenone
25g
C16H24O
1-4-octylphenyl ethanone, p-octylacetophenone, 4'-n-octylacetophenone, 4'-octylacetophenone, ethanone, 1-4-octylphenyl, p-n-octylacetophenone, 4-n-octylacetophenone, acetophenone, 4'-octyl, 1-acetyl-4-octylbenzene, p-octylaceto-phenone
CCCCCCCCC1=CC=C(C=C1)C(=O)C
232.367
232.37
≥97.0% (GC)
White-Yellow
10541-56-7
MFCD00043682
GARQDIVXKVBJFP-UHFFFAOYSA-N
1-(4-octylphenyl)ethanone
66344
Crystal-Lump at 20°C

Chemical Identifiers

10541-56-7
232.367
GARQDIVXKVBJFP-UHFFFAOYSA-N
66344
CCCCCCCCC1=CC=C(C=C1)C(=O)C
C16H24O
MFCD00043682
1-4-octylphenyl ethanone, p-octylacetophenone, 4'-n-octylacetophenone, 4'-octylacetophenone, ethanone, 1-4-octylphenyl, p-n-octylacetophenone, 4-n-octylacetophenone, acetophenone, 4'-octyl, 1-acetyl-4-octylbenzene, p-octylaceto-phenone
1-(4-octylphenyl)ethanone
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3540

TSCA : Yes

Documents