Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Octylacetophenone, 98%

$82.97 - $314.01

Chemical Identifiers

Chemical Name or Material 4'-n-Octylacetophenone
CAS 10541-56-7
Molecular Formula C16H24O
Formula Weight 232.37
MDL Number MFCD00043682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1381703

 
alfa aesar™
A1381703
1g Each for $82.97

AAA1381706

 
alfa aesar™
A1381706
5g Each for $314.01
Description & Specifications

Specifications

Formula Weight 232.37
Chemical Name or Material 4'-n-Octylacetophenone
TSCA TSCA
Boiling Point 152° to 153°C (1mmHg)
Assay 98%
Quantity 1g
Molecular Formula C16H24O
MDL Number MFCD00043682
CAS 10541-56-7