Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester, 97%

$108.20 - $326.32

Chemical Identifiers

Chemical Name or Material 4-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester
CAS 1073353-58-8
Molecular Formula C15H22BNO4
Formula Weight 291.16
MDL Number MFCD09953501
Synonym [4-(N,O-Dimethyl hydroxylaminocarbonyl)phenyl] boronic acid pinacol ester; N,O-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydroxamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2834103

 
alfa aesar™
H2834103
1g Each for $108.20

AAH2834106

 
alfa aesar™
H2834106
5g Each for $326.32
Description & Specifications

Specifications

Formula Weight 291.16
Chemical Name or Material 4-(N,O-Dimethylhydroxylaminocarbonyl)benzeneboronic acid pinacol ester
Melting Point 96° to 98°C
Assay 97%
Quantity 1g
Molecular Formula C15H22BNO4
MDL Number MFCD09953501
CAS 1073353-58-8
Synonym [4-(N,O-Dimethyl hydroxylaminocarbonyl)phenyl] boronic acid pinacol ester; N,O-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzohydroxamate