Learn More
Alfa Aesar™ 4-n-Nonylphenol, 98+%
Manufacturer: Alfa Aesar™ A1560906
Description
Applications4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).
Solubility
Sparingly Soluble (0.020 g/L) at 25°C.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications
4-n-Nonylphenol | |
127°C to 130°C (27mmHg) | |
5g | |
C15H24O | |
UN2430 | |
4-n-nonylphenol, p-nonylphenol, phenol, 4-nonyl, para-nonylphenol, p-n-nonylphenol, phenol, nonyl, phenol, p-nonyl, nonylphenol mixed, phenol, nonyl derivs., 4-nonyl phenol | |
CCCCCCCCCC1=CC=C(C=C1)O | |
220.356 | |
CHEBI:34440 | |
≥98% |
150°C (302°F) | |
43°C to 46°C | |
104-40-5 | |
MFCD00002396 | |
2047450 | |
IGFHQQFPSIBGKE-UHFFFAOYSA-N | |
4-nonylphenol | |
1752 | |
220.36 |
Chemical Identifiers
104-40-5 | |
220.356 | |
IGFHQQFPSIBGKE-UHFFFAOYSA-N | |
1752 | |
4-nonylphenol |
C15H24O | |
MFCD00002396 | |
4-n-nonylphenol, p-nonylphenol, phenol, 4-nonyl, para-nonylphenol, p-n-nonylphenol, phenol, nonyl, phenol, p-nonyl, nonylphenol mixed, phenol, nonyl derivs., 4-nonyl phenol | |
CHEBI:34440 | |
CCCCCCCCCC1=CC=C(C=C1)O |
Safety and Handling
GHS H Statement
H314-H318-H361-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
RTECSNumber : SM5630000
TSCA : TSCA