Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Nonyloxybenzaldehyde, 98%

$53.49 - $201.91

Chemical Identifiers

Chemical Name or Material 4-n-Nonyloxybenzaldehyde
CAS 50262-46-9
Molecular Formula C16H24O2
Formula Weight 248.37
MDL Number MFCD00043712
Beilstein 2529857
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0911004

  SDS
alfa aesar™
L0911004
2g Each for $53.49

AAL0911009

  SDS
alfa aesar™
L0911009
10g Each for $201.91
Description & Specifications

Specifications

Formula Weight 248.37
CAS 50262-46-9
Molecular Formula C16H24O2
Chemical Name or Material 4-n-Nonyloxybenzaldehyde
MDL Number MFCD00043712
Assay 98%
Quantity 2g
Boiling Point 180° to 182°C (4mmHg)
Beilstein 2529857