Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(n-Nonyloxy)benzoic acid, 97%

$23.80 - $68.86

Chemical Identifiers

Chemical Name or Material 4-(n-Nonyloxy)benzoic acid
CAS 15872-43-2
Molecular Formula C16H24O3
Formula Weight 264.37
MDL Number MFCD00043501
Beilstein 2213560
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049603

  SDS
alfa aesar™
B2049603
1g Each for $23.80

AAB2049606

  SDS
alfa aesar™
B2049606
5g Each for $68.86
Description & Specifications

Specifications

Formula Weight 264.37
Melting Point ≈143°C
Beilstein 2213560
Assay 97%
MDL Number MFCD00043501
CAS 15872-43-2
Chemical Name or Material 4-(n-Nonyloxy)benzoic acid
Molecular Formula C16H24O3
Quantity 1g