Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Nonylbiphenyl, 98%

$48.49 - $48.49

Chemical Identifiers

Chemical Name or Material 4-n-Nonylbiphenyl
CAS 93972-01-1
Molecular Formula C21H28
Formula Weight 280.46
MDL Number MFCD00100666
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1740203

 
alfa aesar™
A1740203
1g Each for $48.49
Description & Specifications

Specifications

Formula Weight 280.46
Chemical Name or Material 4-n-Nonylbiphenyl
Assay 98%
Quantity 1g
Molecular Formula C21H28
MDL Number MFCD00100666
CAS 93972-01-1