Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Nonylacetophenone, 97%

$42.34 - $149.18

Chemical Identifiers

Chemical Name or Material 4'-n-Nonylacetophenone
CAS 37593-05-8
Molecular Formula C17H26O
Formula Weight 246.39
MDL Number MFCD00043717
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1283103

  SDS
alfa aesar™
A1283103
1g Each for $42.34

AAA1283106

  SDS
alfa aesar™
A1283106
5g Each for $149.18
Description & Specifications

Specifications

Formula Weight 246.39
MDL Number MFCD00043717
Molecular Formula C17H26O
Quantity 1g
Boiling Point 159° to 161°C (2mmHg)
Chemical Name or Material 4'-n-Nonylacetophenone
Assay 97%
CAS 37593-05-8