Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(N-Nitrosomethylamino)-1-(3-pyridyl)-1-butanone, 98+%

$177.01 - $177.01

Chemical Identifiers

Chemical Name or Material 4-(N-Nitrosomethylamino)-1-(3-pyridyl)-1-butanone
CAS 64091-91-4
Molecular Formula C10H13N3O2
Formula Weight 207.23
MDL Number MFCD00274580
Beilstein 3548355
Synonym NNK
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL14860MB

 
alfa aesar™
L14860MB
25mg Each for $177.01
Description & Specifications

Specifications

Formula Weight 207.23
Chemical Name or Material 4-(N-Nitrosomethylamino)-1-(3-pyridyl)-1-butanone
Molecular Formula C10H13N3O2
Beilstein 3548355
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 63° to 65°C
Assay ≥98%
Quantity 25mg
MDL Number MFCD00274580
CAS 64091-91-4
Synonym NNK
Health Hazard 1 Danger
Recommended Storage −30° to −10°C
RTECS Number OB6465000
Health Hazard 2 GHS H Statement
H350
May cause cancer.
Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350
May cause cancer.

−30° to −10°C