missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[N-Methyl-N-(2-nitrophenyl)sulfamoyl]benzeneboronic acid, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54641MD

 View more versions of this product

Catalog No. AAH54641MD



Specifications

Specifications

4-[N-Methyl-N-(2-nitrophenyl)sulfamoyl]benzeneboronic acid
1449133-61-2
MFCD20265307
LXCUTVNBMYMJPZ-UHFFFAOYSA-N
[4-[methyl-(2-nitrophenyl)sulfamoyl]phenyl]boronic acid
73995905
97%
250mg
C13H13BN2O6S
4-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
B(C1=CC=C(C=C1)S(=O)(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])(O)O
336.125
336.13

Chemical Identifiers

1449133-61-2
336.125
LXCUTVNBMYMJPZ-UHFFFAOYSA-N
73995905
B(C1=CC=C(C=C1)S(=O)(=O)N(C)C2=CC=CC=C2[N+](=O)[O-])(O)O
C13H13BN2O6S
MFCD20265307
4-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
[4-[methyl-(2-nitrophenyl)sulfamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents