Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Hexyloxyphenol, 98%

$70.95 - $949.73

Chemical Identifiers

Chemical Name or Material 4-n-Hexyloxyphenol
CAS 18979-55-0
Molecular Formula C12H18O2
Formula Weight 194.28
MDL Number MFCD00002337
Beilstein 473079
Synonym Hydroquinone monohexyl ether
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1171106

 
alfa aesar™
A1171106
5g Each for $70.95

AAA1171114

 
alfa aesar™
A1171114
25g Each for $289.38

AAA1171122

 
alfa aesar™
A1171122
100g Each for $949.73
Description & Specifications

Specifications

Formula Weight 194.28
Chemical Name or Material 4-n-Hexyloxyphenol
TSCA TSCA
Assay 98%
Molecular Formula C12H18O2
MDL Number MFCD00002337
CAS 18979-55-0
Beilstein 473079
Synonym Hydroquinone monohexyl ether
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 44° to 47°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.