missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Hexyloxyaniline, 99%

Manufacturer:  Alfa Aesar™ H5586814

 View more versions of this product

Catalog No. AAH5586814



Specifications

Specifications

4-n-Hexyloxyaniline
White to Brown
25g
C12H19NO
4-hexyloxy aniline
CCCCCCOC1=CC=C(C=C1)N
193.29
193.29
99%
155°C to 158°C (5mmHg)
43°C to 45°C
39905-57-2
MFCD00007868
DJRKHTCUXRGYEU-UHFFFAOYSA-N
4-hexoxyaniline
38360
Solid

Chemical Identifiers

39905-57-2
193.29
DJRKHTCUXRGYEU-UHFFFAOYSA-N
38360
CCCCCCOC1=CC=C(C=C1)N
C12H19NO
MFCD00007868
4-hexyloxy aniline
4-hexoxyaniline
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BY2308000

TSCA : TSCA

Documents