Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Hexylbiphenyl, 98%

$50.34 - $174.78

Chemical Identifiers

Chemical Name or Material 4-n-Hexylbiphenyl
CAS 59662-31-6
Molecular Formula C18H22
Formula Weight 238.37
MDL Number MFCD00009527
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1680103

 
alfa aesar™
A1680103
1g Each for $50.34

AAA1680106

 
alfa aesar™
A1680106
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 238.37
Melting Point 29° to 31°C
Boiling Point 148° to 150°C (2mmHg)
Quantity 1g
Flash Point >110°C (230°F)
CAS 59662-31-6
Molecular Formula C18H22
MDL Number MFCD00009527
Chemical Name or Material 4-n-Hexylbiphenyl
Assay 98%