missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Hexylbenzeneboronic acid, 97%

$67.70 - $214.35

Chemical Identifiers

CAS 105365-50-2
Molecular Formula C12H19BO2
Molecular Weight (g/mol) 206.092
MDL Number MFCD04039029
InChI Key CFYBKGAFWFRFRJ-UHFFFAOYSA-N
Synonym 4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
PubChem CID 4197864
IUPAC Name (4-hexylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCCCC)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64555MD View Documents Alfa Aesar™
H64555MD
250mg Each for $67.70
 
AAH6455503 View Documents Alfa Aesar™
H6455503
1g Each for $214.35
 
Specifications

Chemical Identifiers

105365-50-2
206.092
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
4197864
B(C1=CC=C(C=C1)CCCCCC)(O)O
C12H19BO2
MFCD04039029
4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
(4-hexylphenyl)boronic acid

Specifications

250mg
C12H19BO2
4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid, boronic acid, 4-hexylphenyl, 4-hexylphenylboronicacid, p-hexylphenylboronic acid, 4-hexylbenzeneboronic acid, 4-hexylphenyl-boronic acid
B(C1=CC=C(C=C1)CCCCCC)(O)O
206.092
206.09
97%
105365-50-2
MFCD04039029
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
(4-hexylphenyl)boronic acid
4197864
Solid
4-n-Hexylbenzeneboronic acid