Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Hexylacetophenone, 97%

$62.34 - $247.82

Chemical Identifiers

Chemical Name or Material 4'-n-Hexylacetophenone
CAS 37592-72-6
Molecular Formula C14H20O
Formula Weight 204.31
MDL Number MFCD00043677
Beilstein 1942064
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0897304

 
alfa aesar™
L0897304
2g Each for $62.34

AAL0897309

 
alfa aesar™
L0897309
10g Each for $247.82
Description & Specifications

Specifications

Formula Weight 204.31
Assay 97%
Molecular Formula C14H20O
MDL Number MFCD00043677
CAS 37592-72-6
Beilstein 1942064
Flash Point >110°C (230°F)
Boiling Point 132° to 135°C (2mmHg)
Quantity 2g
Chemical Name or Material 4'-n-Hexylacetophenone
TSCA TSCA