Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Hexadecyloxyaniline, 98%

$81.22 - $304.36

Chemical Identifiers

Chemical Name or Material 4-n-Hexadecyloxyaniline
CAS 7502-06-9
Molecular Formula C22H39NO
Formula Weight 333.56
MDL Number MFCD00025325
Beilstein 2655373
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0897004

  SDS
alfa aesar™
L0897004
2g Each for $81.22

AAL0897009

  SDS
alfa aesar™
L0897009
10g Each for $304.36
Description & Specifications

Specifications

Formula Weight 333.56
CAS 7502-06-9
Health Hazard 1 Danger
Chemical Name or Material 4-n-Hexadecyloxyaniline
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Molecular Formula C22H39NO
MDL Number MFCD00025325
Assay 98%
Beilstein 2655373
Quantity 2g
Melting Point 59° to 61°C
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.