Please login to your online account to display your discounted pricing

4-n-Heptyloxybenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-n-Heptyloxybenzoic acid
Name Note 98%
CAS 15872-42-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H20O3
Linear Formula H3C(CH)6OC6H4COOH
Formula Weight 236.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC338320100

  SDS
thermo fisher scientific
338320100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 236.31
Quantity 10g
Color White
EINECS Number 239-995-3
CAS Max % 100.0
Chemical Name or Material 4-n-Heptyloxybenzoic acid
Name Note 98%
Infrared Spectrum Authentic
Molecular Formula C14H20O3
Packaging Glass bottle
Linear Formula H3C(CH)6OC6H4COOH
MDL Number MFCD00013992
CAS 15872-42-1
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.