Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Heptyloxybenzoic acid, 98%

$78.80 - $142.08

Chemical Identifiers

Chemical Name or Material 4-n-Heptyloxybenzoic acid
CAS 15872-42-1
Molecular Formula C14H20O3
Formula Weight 236.31
MDL Number MFCD00013992
Beilstein 2211867
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1405909

 
alfa aesar™
A1405909
10g Each for $78.80

AAA1405914

 
alfa aesar™
A1405914
25g Each for $142.08
Description & Specifications

Specifications

Formula Weight 236.31
Chemical Name or Material 4-n-Heptyloxybenzoic acid
Assay 98%
Molecular Formula C14H20O3
MDL Number MFCD00013992
CAS 15872-42-1
Beilstein 2211867
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 92° to 94°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.