Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-n-Heptyloxyaniline, 98%

$84.82 - $292.31

Chemical Identifiers

Chemical Name or Material 4-n-Heptyloxyaniline
CAS 39905-44-7
Molecular Formula C13H21NO
Formula Weight 207.32
MDL Number MFCD00038135
Beilstein 2937583
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0896204

  SDS
alfa aesar™
L0896204
2g Each for $84.82

AAL0896209

  SDS
alfa aesar™
L0896209
10g Each for $292.31
Description & Specifications

Specifications

Formula Weight 207.32
MDL Number MFCD00038135
Health Hazard 1 Danger
CAS 39905-44-7
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Chemical Name or Material 4-n-Heptyloxyaniline
Quantity 2g
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H21NO
Boiling Point 145° to 147°C (0.5mmHg)
Beilstein 2937583
Melting Point 44° to 46°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.