missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-n-Heptylbenzoic acid, 99+%, Thermo Scientific Chemicals
$111.48 - $345.96
Chemical Identifiers
CAS | 38350-87-7 |
---|---|
Molecular Formula | C14H20O2 |
Molecular Weight (g/mol) | 220.312 |
MDL Number | MFCD00009722 |
InChI Key | VSUKEWPHURLYTK-UHFFFAOYSA-N |
Synonym | benzoic acid, 4-heptyl, 4-n-heptylbenzoic acid, 4-heptylbenzoicacid, 4-heptylbenzoate, p-heptylbenzoic acid, acmc-209izm, 4-n-heptyl-benzoic acid, 4-heptylbenzoic acid |
PubChem CID | 170036 |
IUPAC Name | 4-heptylbenzoic acid |
SMILES | CCCCCCCC1=CC=C(C=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2258906
|
Thermo Scientific Chemicals
B2258906 |
5 g |
Each for $111.48
|
|
|||||
AAB2258914
|
Thermo Scientific Chemicals
B2258914 |
25 g |
Each for $345.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
38350-87-7 | |
220.312 | |
VSUKEWPHURLYTK-UHFFFAOYSA-N | |
170036 | |
CCCCCCCC1=CC=C(C=C1)C(=O)O |
C14H20O2 | |
MFCD00009722 | |
benzoic acid, 4-heptyl, 4-n-heptylbenzoic acid, 4-heptylbenzoicacid, 4-heptylbenzoate, p-heptylbenzoic acid, acmc-209izm, 4-n-heptyl-benzoic acid, 4-heptylbenzoic acid | |
4-heptylbenzoic acid |
Specifications
38350-87-7 | |
C14H20O2 | |
5 g | |
benzoic acid, 4-heptyl, 4-n-heptylbenzoic acid, 4-heptylbenzoicacid, 4-heptylbenzoate, p-heptylbenzoic acid, acmc-209izm, 4-n-heptyl-benzoic acid, 4-heptylbenzoic acid | |
CCCCCCCC1=CC=C(C=C1)C(=O)O | |
220.312 | |
220.31 | |
4-n-Heptylbenzoic acid |
101°C to 102°C | |
MFCD00009722 | |
2047135 | |
VSUKEWPHURLYTK-UHFFFAOYSA-N | |
4-heptylbenzoic acid | |
170036 | |
≥99% |
Safety and Handling
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302-H319-H335
EINECSNumber : 253-894-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only