Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-n-Heptylacetophenone, 97%

$40.03 - $148.85

Chemical Identifiers

Chemical Name or Material 4'-n-Heptylacetophenone
CAS 37593-03-6
Molecular Formula C15H22O
Formula Weight 218.34
MDL Number MFCD00038342
Beilstein 1866760
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0895903

 
alfa aesar™
L0895903
1g Each for $40.03

AAL0895906

 
alfa aesar™
L0895906
5g Each for $148.85
Description & Specifications

Specifications

Formula Weight 218.34
Chemical Name or Material 4'-n-Heptylacetophenone
Assay 97%
Molecular Formula C15H22O
MDL Number MFCD00038342
CAS 37593-03-6
TSCA TSCA
Beilstein 1866760
Flash Point >110°C (230°F)
Boiling Point 155° to 157°C (2mmHg)
Quantity 1g